missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 14 gal.
Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.
Supplier: Eagle™ 1610MBKBG1
Description
- Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
- For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
- Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
- If used with leaking materials, drums must be packed with sufficient sorbents
- No free liquids
Specifications
Drum | |
Metal Lever-lock | |
53.06 L | |
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration | |
Open | |
63.5 cm | |
10 lb. |
Yes | |
14 gal. | |
Black | |
HDPE | |
25 in. | |
0.75 in., 2 in. (Bung Holes) |