missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eagle™ Lab Pack Open Head Poly Drum with Bung Holes, 55 gal.

Drum is constructed of blow-molded high-density polyethylene (HDPE) with UV inhibitors, is durable and lightweight, as well as weather and chemical resistant. Bung lid includes two holes (3/4 in. and 2 in. diameters) and is ideal for safe storage, as well as handling or transportation of hazardous materials.

Supplier:  Eagle™ 1655MBLKBG

 View more versions of this product

Catalog No. 19338413


Explore available promotions

Description

Description

  • Constructed from blow-molded high-density polyethylene (HDPE) with UV inhibitors
  • For use with smaller non-damaged bottles, cans, carboys and 5 gal. pails of non-leaking hazardous materials
  • Bung lid includes two holes, one of each diameter: 0.75 in. and 2 in.
  • If used with leaking materials, drums must be packed with sufficient sorbents
  • No free liquids
Specifications

Specifications

Drum
Metal Lever-lock
208.45 L
Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Open
98 cm
23.5 lb.
Yes
55 gal.
Black
HDPE
38.6 in.
0.75 in., 2 in. (Bung Holes)
SDS
Documents

Documents

Promotions

Promotions