Learn More
Labconco™ Anti-Static Ionizer Fan
Circulate ionized air inside Labconco Glove Boxes.
Supplier: Labconco™ 5234401
Description
Anti-Static Ionizer Fan circulates ionized air inside Labconco Protector and Precise Glove Boxes and XPert Weigh Boxes to reduce static charge. Fan requires installation.Specifications
| Protector Combination Glove Box | |
| 1.6 Kg | |
| Circulates ionized air inside Glove Box to reduce Static Charge |
| 3.5 lb. | |
| 230 V |