Labconco™ Bottle Holder
Attaches to rear baffle perforations
Supplier:  Labconco™ 3927700
Catalog No. 16-108-398
Explore available promotions
Description
Description
  • Powder-coated steel construction
  • Fits the XPert™ Balance Enclosure, Station or System for 2 ft. models; Protector™ XStream™ Laboratory Hood for 2 ft. models; Protector XVS™ Ventilation Station; Purifier™ Class I for 2 ft. models and HEPA Filtered Enclosures (manufactured after July 2004); and Protector Work Station for 2 ft. models (manufactured after August 2004)
  • 3 in. dia. (7.6cm) opening to hold most standard squirt bottles
Specifications
Specifications
1 lb.
0.5 Kg
Epoxy-coated Steel