Labconco™ Crude Fiber Apparatus
Six 350-watt heaters for 600mL beakers
Supplier:  Labconco™ 3000100
Catalog No. 09-681B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Includes: 350-Watt heaters, 6; Infinite heat controls, 6; Tall-form 600mL borosilicate glass beakers—six for test and six spares, 12; Stainless-steel condensers, 6
Description
Description
Benchtop unit for determining crude fiber in feed, food and other agricultural products as outlined in AOAC™ methods. Amount of crude fiber is determined by subjecting sample to simulated digestive action. Samples are boiled in acid, washed, boiled in alkali and washed again. Loss upon ignition is defined as crude fiber.
  • Compression spring holds beaker rigidly between heater and condenser during digestion
  • Tight seal keeps acid and alkali at constant volumes during boiling
  • Complete for six samples
  • Requires connection to water source
Order Info
Dimensions: 9L x 35.68W x 27.18 in.H (23 x 91 x 69cm). Shipping weight: 137 lb. (62kg). Base cabinet not included. For 115V 50/60Hz, 2100w. Must be wired directly to line (line cord not supplied).
Specifications
Specifications
Crude Fiber Apparatus
50/60 Hz
350 W
23 cm
Determination of crude fiber in feed, food and other agricultural products
69 cm
1 Ea.
18.3 A
115 V
9 in.
115 V, 50/60 Hz
27.2 in.
Six 350w heaters, six infinite heat controls, six tall-form 600ml borosilicate glass beakers for test and six spares, six stainless steel condensers