Labconco™ Distillation Grid Kits
Accessory for fume hoods
Supplier:  Labconco™ 9725200
Catalog No. 09-039-071
Explore available promotions
Includes: Stainless-steel rods, connectors, instruction sheet and drill template required for installation
Description
Description
  • For use with Protector Premier, XStream, XL Benchtop or XLE Laboratory Hood
Specifications
Specifications
Stainless steel rods, connectors, instruction sheet and drill template for installation
Distillation Grid Kit
Protector Premier, Xstream, XL benchtop or XLE laboratory hood