Labconco™ Kjeldahl Digestion Tubes for Rapid Digestor-4 and Rapid Digestor-25
For Rapid Digestor-25 and Rapid Digestor-4
Supplier:  Labconco™ 2304025
 View more versions of this product
Catalog No. 13-162B
Explore available promotions
Description
Description
Digestion Tubes
  • 250mL tubes are made of borosilicate glass
  • Heavy-duty retention lip and contoured bottom for optimum heat transfer and minimal thermal shock
  • Provide uniformity, impact strength and chemical resistance
  • Volumetric tubes have easy-to-read calibration mark and built-in constriction for maximum refluxing and digestion speed
Specifications
Specifications
Straight Digestion Tube
250 mL
Borosilicate Glass
8.45 oz.
Rapid digestor-25 and rapid digestor-4
25 Pk.