Labconco™ Lyph-Lock™ Freeze-Dry Flask Assembly with standard taper 19/38 Joints
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring
Supplier: Labconco™ 7550400
Description
A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring
Flasks should be filled no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is assured. Adapters are required to attach flasks to freeze dry valve ports (sold separately).
Specifications
2.32 in. | |
1.96 in. | |
Glass top and bottom, Silicone Rubber ring seal | |
3 lb. |
100 mL | |
5 cm | |
19/38 | |
5.9 cm |