missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Precise™ HEPA-Filtered Glove Boxes
Provide a leak-tight physical barrier to protect operator from hazardous airborne particulates and powders
Supplier: Labconco™ 5220200
Includes:
- Integral neoprene gloves (size 9.75, 30 in. [76cm] long, 15mil gauge)
- 22w fluorescent lamp
- Interior ceiling-mounted electrical duplex receptacle with 5A (115V model)
- Built-in blower with speed control capable of airflow from 13 to 100cfm
- Switches for light, blower, and innermost electrical receptacle
- Inlet and outlet HEPA filters and test port for challenging HEPA filter
- Main chamber static pressure gauge to monitor HEPA filter loading integrity
Requires : Supporting base (10-358-108, -109; order separately)
Description
Leak-tight design and HEPA inlet and outlet filters make this box suitable for pharmaceutical research, nanotechnology and biochemistry. Also suitable for work with microorganisms, low-level radiochemicals, chemical carcinogens and asbestos, as well as cleanroom applications requiring ISO Class 3 conditions.
Design Features
- One-piece molded, medium-density polyethylene shell
- Dry powder epoxy-coated steel superstructure and interior shelf of transfer chamber
- Approx. 13 cu. ft. main chamber
- 0.25 in. thick (0.64cm) laminated safety glass viewing window with powder-coated steel frame and neoprene gasket; cut-out is 26.37 x 12.5 in. (67.1 x 31.8cm)
- Upward-pivoting inner and outer transfer chamber doors counterbalanced and equipped with quick-latches
- Black, chemical-resistant phenolic core work surface sealed to glove box floor
- Dual HEPA filters boast 99.99% efficiency against particulates down to 0.3 microns
- 0.05ppm or lower average tracer gas levels detected during perimeter scan testing surpass ASHRAE™ 110 standards
- Factory testing with mass spectrometer reveals no detectable leaks greater than 1 x 10-3mL/sec. when tested while glove box is pressurized with helium at 1 in. water gauge
- Vibration is less than 1.5 x 10-5 inches displacement when measured at work surface at both minimum and maximum airflow volumes
Order Info
- Overall dimensions: 31.62L x 52.75W x 40 in.H (80 x 134 x 102cm)
- Main chamber interior measures 27.5D x 33.5W x 25 in.H (69.9 x 85.1 x 63.5cm)
- Transfer chamber is 11 in. nominal I.D. x 12 in.L (28 x 30cm)
- Shipping weight: 280 lb. (127kg)
Certifications
ETL listed
Specifications
Glove Box | |
8 A | |
31.6 in. | |
80 cm | |
Pharmaceutical research, nanotechnology and biochemistry | |
25 in. | |
60 Hz | |
2 | |
280 lb. | |
57.2 in. | |
147 cm |
HEPA | |
UL, ETL, CAN/CSA | |
27.5 in. | |
70 cm | |
40 in. | |
102 cm | |
Polyethylene | |
115 V | |
127 kg | |
33.5 in. | |
85.09 cm |