Labconco™ PrimeMate™ Vacuum Pump Oil
For use with the Labconco PrimeMate Oil Change System
Supplier: Labconco™ 1988001
Description
- Choice of one gallon (3.8L) hydro-treated paraffinic hydrocarbon oil or synthetic hydrocarbon oil
- Recommended for Labconco general purpose vacuum pumps
Specifications
Vacuum Pump Oil | |
PrimeMate Oil Change System | |
3.8 L |