missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Protector™ Standard Storage Cabinet
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
Supplier: Labconco™ 9900800
Includes:
Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in.
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Order Info
Work surface and interior shelf kits are sold separately
Specifications
Storage Cabinet | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
12 in. | |
Left Hinge, Manual Close | |
55.88 cm | |
30.48 cm | |
1 | |
90.17 to 93.34 cm |
22 in. | |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. | |
29.48 kg | |
65 lb. | |
362.8 kg | |
35.5 to 36.75 in. | |
800 lb. | |
Epoxy-coated Steel |