missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Labconco™ Protector™ Standard Storage Cabinet
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
Supplier: Labconco™ 9900600
Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Order Info
Work surface and interior shelf kits are sold separately
Specifications
22 in. | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
35.5 to 36.75 in. | |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. | |
58.96 kg | |
130 lb. | |
60.96 cm | |
800 lb. | |
Epoxy-coated Steel |
Storage Cabinet | |
90.17 to 93.34 cm | |
Epoxy | |
24 in. | |
Left Hinge, Manual Close | |
362.87 kg | |
1 | |
White |