missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ Protector™ Standard Storage Cabinet

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

Supplier:  Labconco™ 9900100

 View more versions of this product

Catalog No. 16-305-79


Explore available promotions

Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .

Description

Description

The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.

  • Support loads up to 800 lb. (363kg)
  • Durable epoxy-coated steel construction
  • Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
  • Manual-closing, non-locking door(s)

Order Info

Work surface and interior shelf kits are sold separately

Specifications

Specifications

22 in.
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent
35.5 to 36.75 in.
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in.
81.64 kg
180 lb.
91.44 cm
800 lb.
Epoxy-coated Steel
Storage Cabinet
90.17 to 93.34 cm
Epoxy
36 in.
Manual Close
362.87 kg
2
White
Videos
SDS
Documents

Documents

Promotions

Promotions