missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Labconco™ SteamScrubber™ Glassware Washer Accessory, Upper Spindle Rack

Hold your laboratory glassware securely and get it extra clean with a Labconco Upper Spindle Rack. This SteamScrubber accessory provides direct-injection cleaning right through the spindles.

Supplier:  Labconco™ 4595800

Catalog No. 10-359-201


Explore available promotions

Includes: Slide release latches, 30 removable spindles, 30 adjustable height clips, 10 small spindles, 15 plastic glassware holders, tool to remove the spindles, 30 nylon plugs, wash arm

Requires : SteamScrubber with upper slide arms installed

Description

Description

  • Constructed of Type 304 stainless steel
  • Spindles may be removed and plugged to accommodate accessory inserts (sold separately)
  • Spindle dimensions: 5.25 in. high x 1/4 in. OD
  • Accommodates glassware up to 8.5 in. high.
Specifications

Specifications

Stainless Steel (304)
30
53 x 51 x 18cm (20.9 x 20 x 7.2 in.)
7.25 kg
Rack
SteamScrubber™ Glassware Washer
16 lb.
Videos
SDS
Documents

Documents

Promotions

Promotions