missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Label Printer Hard Case
Protect, transport and store your M511 Label Printer, plus printer accessories and label cartridges.
Supplier: Brady™ M511HC
Description
Protect, transport, and store your M511 Label Printer, plus printer accessories and label cartridges.
- Protects your printer from impacts and dust.
- Keeps printer, accessories and label cartridges organized and on hand.
- Durable hard case allows for easy mobility.
Specifications
Hard Case | |
Brady Label Printers | |
6.5 in. | |
17.5 in. | |
(1) Hard Case |
Blue | |
Plastic | |
17 in. | |
M511 Printer Hard Case | |
Asset Tracking;Circuit Board Labeling;Component and Equipment Labeling;Data and Telecommunications Labeling;Electrical Labeling;Facility Identification;General Identification;Inventory and Inspection Labeling;Laboratory Labeling;Lean and 5S Labeling;Safety Identification;Warehouse Marking;Wire and Cable Labeling |