Fisherbrand™ Laboratory Ball Valve
Laboratory Ball Valve is deck mounted.
Description
- Chrome plated brass
- Quarter-turn open/close
- Forged brass lever with color-coded index disc
Specifications
Chrome | |
Chrome-plated Brass | |
Ball valve assembly | |
5.7 in. | |
1.6 in. | |
3.6 in. |
Blue | |
1 Turret, Gas Service | |
3/8 in. IPS mounting shank with a 3/8 in. NPT male inlet | |
14.5 cm | |
4.1 cm | |
9.2 cm |