Fisherbrand™ Laboratory Ball Valve
Laboratory Ball Valve is deck mounted.
 View more versions of this product
Catalog No. 06000405
Explore available promotions
Description
Description
  • Chrome plated brass
  • Quarter-turn open/close
  • Forged brass lever with color-coded index disc
Specifications
Specifications
Chrome
Chrome-plated Brass
Ball valve assembly
5.7 in.
1.6 in.
3.6 in.
Yellow
1 Turret, Vacuum Service
3/8 in. IPS mounting shank with a 3/8 in. NPT male inlet
14.5 cm
4.1 cm
9.2 cm