Fisherbrand™ Laboratory Ball Valve
Laboratory Ball Valve is deck mounted.
Description
- Chrome plated brass
- Quarter-turn open/close
- Forged brass lever with color-coded index disc
Specifications
Chrome | |
Chrome-plated Brass | |
Ball valve assembly | |
24.8 cm | |
3.6 in. |
Blue | |
4 Turrets, Gas Service | |
9.8 in. | |
9.8 in. | |
9.2 cm |