missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Heidolph Laboratory Line Autoclaves
Designed to meet todayâs changing lab sterilization needs, these autoclaves offer a steam generator, vacuum pump, and cooling coils often found on larger autoclaves in a compact platform
Supplier: Heidolph 023210683
Description
- Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
- For use with Agar preparation as well as the sterilization of liquids, pipettes, glasses, instruments, and biohazards with various cooling options
- Equipped with a vacuum pump, cooling coils, and a steam generator for all laboratory sterilization needs
- Compact size allows autoclave to fit on a standard benchtop
- Fast cooling (C) saves up to 75% of the cool down times of liquid samples
- Immediate Steam (G) and Efficient Air Removal (PV) Steam generator in combination with vacuum (PV G) for complete drying
- PID (Proportional Integral Differential) pressure control
- Stores the last 200 cycles in built-in memory
- Two independent, flexible PT100 temperature sensors prevent boil over of liquids and explosions of bottles
- Compressed air is passed through a microbiological filter into the autoclave chamber after sterilization to prevent a drop in pressure which causes load deformation, cracks, and spills
- Cold water is circulated through cooling pipes to rapidly reduce the temperature of the chamber and liquid load to a safe temperature
- Highly efficient drying is achieved by uniformly heating the chamber wall of the benchtop autoclave using a coiled pipe
- Guarantees that even the most difficult loads will dry such as textiles, porous loads, hollow instruments, and tips
- The vacuum pump can be used for post vacuum drying to ensure improved drying of porous loads and hollow instruments such as pipette tips
- Equipped with a heating plate attached under the chamber for heating during the drying phase
- Double locking safety device prevents door from opening while chamber is pressurized
- Control lock-out switch prevents starting a cycle if door is not properly locked
- Control system protection prevents door from opening at high pressure and temperature
Specifications
Not Autoclavable | |
41.5 cm | |
7 gal. | |
45 cm | |
18 in. | |
Sterilization | |
29.9 in. | |
44 cm | |
2840 EL-PV-GC | |
221°F to 278°F | |
Multi-color to display the stage of the cycle | |
43 kg | |
230 V 1 Phase | |
6.7 in. | |
Non-sterile | |
20.9 x 24.8 x 17.3 in. (53 x 63 x 44 cm) |
â½16 A | |
16.3 in. | |
28 L | |
11 in. | |
28 cm | |
Liquids, Glassware, Utensils, Wrapped, Unwrapped, and Solids | |
17.3 in. | |
76 cm | |
Benchtop Autoclave | |
105°C to 137°C | |
94.8 lb. | |
Included | |
2200 w | |
17 cm | |
76.2 kg |