missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Laboratory Notebooks

Space for dates and signatures, record-keeping instructions, table of contents and more.

Supplier:  Thermo Scientific™ 63014000E

 View more versions of this product

Catalog No. S40527



Explore available promotions

Description

Description

  • Sewn-in paper pages
  • Waterproof polyethylene cover
  • Size: 8.5 × 11 in. (21.6 × 27.9cm)
  • Page count: 100

Specifications

Specifications

Polyethylene
8.75 in.
28.57 cm
0.20 in. (5 mm)
Permanently sewn-in paper pages
A4
11.25 in.
Burgundy
28.57 cm
96
Laboratory Notebook
Videos
SDS
Documents

Documents

Promotions

Promotions