Kinematica Labstar Basic Lab Benchtop Autoclave
Kinematica Labstar Basic Compact Lab Benchtop Autoclave with angular chamber, touch screen, password protected. Recommended for the sterilization of simple materials or liquids without removal when dry.
$36650.79 - $103559.91
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance CE Compliant
For Use With (Application) Steam Sterilization of Lab Equipment and Waste
For Use With (Equipment) Glass, Stainless Steel, Cloth, Plastic, Clothing, Biohazardous Materials, Agar, Media
Model Labstar
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Capacity (English) Dimensions (L x W x H) Weight (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Capacity (English) Dimensions (L x W x H) Weight (Metric) Price Quantity  
08451072
View Documents
Kinematica
A1200101
25 L 6.6 gal. 450 x 520 x 660 mm 45 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
08451073
View Documents
Kinematica
A1220101
40 L 11 gal. 620 x 532 x 570 mm 65 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
08451075
View Documents
Kinematica
A1280101
200 L 53 gal. 1250 x 750 x 900 mm 165 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Easy-to-use touchscreen
  • 10 variable program channels for maximum flexibility
  • Password-protected parameters
  • Round chamber (Labstar 200), Angular (Labstar 25, 40, 70)
  • Maximum allowable operating temperature up to 140°C and maximum pressure at 3.8 bar absolute
  • Equipped with electronic pressure sensor and PT100 temp sensor
  • Hinged door with locking mechanism powered by an electric motor
  • Sterilization chamber and door made with stainless steel
  • Housing made with brushed stainless steel
Specifications
Specifications
Not Autoclavable
Steam Sterilization of Lab Equipment and Waste
Labstar
140°C
Easy-To-Use Touchscreen, Password-Protected, 10 Variable Program Channels for Maximum Flexibility, Angular Chamber, Sterilization Chamber and Door Made of Stainless Steel 1.4404/AiSi316L, Equipped with Electronic Pressure Sensor and PT100 Temp Sensor, Hinged Door with Locking Mechanism Powered by an Electric Motor.
60 Hz
Yes
Programmable
Steam Sterilization
CE Compliant
Glass, Stainless Steel, Cloth, Plastic, Clothing, Biohazardous Materials, Agar, Media
Autoclave
Touch Display
Free Promotional Printer (Limited Time)
Stainless Steel
3.8 bar
1 Ea.
Sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'