Learn More
Thermo Scientific™ Labtainer™ 2D BioProcess Container (BPC), Aegis™ 5-14 film, two ports, Luer locks + MPC
Thermo Scientific Labtainer 2D BioProcess Containers (BPCs) are composed of Aegis 5-14 single-web, multi-layer film.
$623.16 - $763.17
Specifications
For Use With (Application) | Bioprocessing |
---|---|
Film Description | Aegis 5-14 |
Port Detail | 3 ports |
Tubing Style | C-Flex |
Certifications/Compliance | ISO 7 |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
SH3104811
|
Thermo Scientific™
SH3104811 |
50 mL |
N/A
|
|
|||||
SH3104812
|
Thermo Scientific™
SH3104812 |
100 mL |
N/A
|
|
|||||
SH3104813
|
Thermo Scientific™
SH3104813 |
250 mL |
N/A
|
|
|||||
SH3104814
|
Thermo Scientific™
SH3104814 |
500 mL |
N/A
|
|
|||||
SH3104815
|
Thermo Scientific™
SH3104815 |
1 L |
N/A
|
|
|||||
SH3104816
|
Thermo Scientific™
SH3104816 |
2 L |
N/A
|
|
|||||
Description
Features of the Labtainer 2D BPC include:
• Helps eliminate post-use cleaning steps required with re-useable containers and reduce cross-contamination risks
• Constructed in an ISO 7 cleanroom under cGMP conditions
Applications of the Labtainer 2D BPC include:
• Harvest from, feed into, and collect samples from bioreactors and fermentors
• Prepare and store buffer, culture media, and process liquid
• Collect chromatography feed and fractions
• Harvest, storage, and transport bulk drug product and bulk drug precursors
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library with over 20,000 unique components that include:
• Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
• Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
• Filters—used for bioburden reduction; sterilization grade filters and vent filters
• Tubing—thermoplastic elastomers, platinum-cured silicone
• Sample ports—septum-style and needle-free ports
• Clamps—sliders, pinch, and crimp bands
Specifications
Bioprocessing | |
3 ports | |
ISO 7 |
Aegis 5-14 | |
C-Flex | |
Sterile |