Learn More
Thermo Scientific™ Labtainer™ BioProcess Container (BPC), 100mL with 2 Ports, Luer Lock, 2D BPC
Simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.
$491.69 - $699.19
Specifications
Capacity (Metric) | 100 mL |
---|---|
Dimensions | 14.7 cm x 14.2 cm (5.8 in. x 5.6 in.) |
Outer Packaging | Packages placed in strong cardboard cartons with product and lot identification |
Protective Inner Packaging | Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area |
Quantity | 10 per case |
Catalog Number | Mfr. No. | Capacity (Metric) | Tubing Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Tubing Style | Price | Quantity | ||||
SH3065812
|
Thermo Scientific™
SH3065812 |
100 mL | 12 in. of 1/4 in. Tubing |
Pack of 10 Each for $491.69
|
|
||||
SH3065712
|
Thermo Scientific™
SH3065712 |
100 mL | No Tubing |
Pack of 10 Each for $699.19
|
|
||||
Description
Using Thermo Scientific™ Labtainer™ BPC bag with 2 top ports (luer lock and MPC insert), simplify cleaning and disposal and reduce cross-contamination risks for 50 to 2000mL volumes of media, buffers, water for injection (WFI) and biological products.
Labtainers are constructed with our CX5-14 single web, multi-layer cast film manufactured in a cGMP facility. Our standard Labtainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function Labtainers can be customized using the industry's largest component library.Highlights:
- 11 sizes from 50mL to 50L
- Film designed to incorporate best combination of flexibility and durability for collection, storage and transport of sterile bioprocess liquids: superior stress resistance, puncture resistance, and physical properties to ensure stability of contents
- Gamma-irradiation sterilized
- Convenient end-port design with a handle at the opposing end
- Space-efficient, ergonomic alternative to rigid walled carboys
- Customizable via the industry's largest library of qualified components
Recommended for:
Sampling – collection of in-process samples at a variety of bioprocessing stages
Harvest – collection and storage of bulk product from bioreactors
Storage – sterile storage of media, buffers and process intermediates
Separation – feeding and receiving of sterile liquids from separation systems
Purification – buffer feeding to purification system, fraction collection and storage
Filling – sterile reservoir for filling systems
Transport & Shipping - bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
100 mL | |
Packages placed in strong cardboard cartons with product and lot identification | |
10 per case | |
Labtainer™ | |
CX5-14 |
14.7 cm x 14.2 cm (5.8 in. x 5.6 in.) | |
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
Gamma Irradiated | |
Edge Ports: 2 x 0.25 in. |