missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lactic Acid (85% w/w/Certified ACS), Fisher Chemical™

$684.23 - $684.23

Chemical Identifiers

CAS 50-21-5
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00004520
InChI Key JVTAAEKCZFNVCJ-UHFFFAOYNA-N
Synonym lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
PubChem CID 612
ChEBI CHEBI:78320
IUPAC Name 2-hydroxypropanoic acid
SMILES CC(O)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
A162-1
SDS View Documents Promotion Details
Thermo Fisher Scientific
BPA1621
1 L Glass Bottle
Each for $684.23
Case of 6 Each for $4,105.38
 
Description

Description

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

Specifications

Chemical Identifiers

50-21-5
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
612
2-hydroxypropanoic acid
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CHEBI:78320
CC(O)C(O)=O

Specifications

2,8
50-21-5,97-73-4
85.0 to 90.0%
CH3CH(OH)COOH
3265
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
612
85.0 to 90.0%
1.200g/cm³
0.020% max.
Viscous Liquid
18°C
122°C
C3H6O3
MFCD00004520
lactic acid, dl-lactic acid, 2-hydroxypropionic acid, milk acid, lactate, polylactic acid, ethylidenelactic acid, lactovagan, tonsillosan, acidum lacticum
CC(O)C(O)=O
90.08
CHEBI:78320
Certified ACS
1 L
Glass Bottle
Lactic Acid, 85% (w/w)
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 8, : Corrosive

SDS
missing translation for 'documents'

missing translation for 'documents'