missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lactic Acid, ACS, 85-90%, Spectrum™ Chemical
$217.88 - $17447.35
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18611178
|
Spectrum Chemical Mfg Cor
L1005125ML |
125 mL | Poly Bottle |
Each for $217.88
|
|
||||
18611181
|
Spectrum Chemical Mfg Cor
L1005500MLPL |
500 mL | Poly Bottle |
Each for $590.35
|
|
||||
18611179
|
Spectrum Chemical Mfg Cor
L100525LT |
2.5 L | Glass Bottle |
Each for $1,035.38
|
|
||||
18611176
|
Spectrum Chemical Mfg Cor
L10054LTPL |
4 L | Poly Bottle |
Each for $1,877.53
|
|
||||
18611177
|
Spectrum Chemical Mfg Cor
L100520LTPL |
20 L | Poly Drum |
Each for $4,599.79
|
|
||||
18611180
|
Spectrum Chemical Mfg Cor
L1005200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
50-21-5 |
Specifications
50-21-5 | |
85 to 90% | |
125 mL | |
0.0002 |
1 | |
ACS | |
Poly Bottle |
CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.