missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lactic Acid, Racemic, USP, 88-92%, Spectrum™ Chemical

L1010, 50-21-5, C3H6O3

Supplier:  Spectrum Chemical Mfg Cor L10104LTGL

 View more versions of this product

Catalog No. 18603682


Explore available promotions

Description

Description

Spectrum™ Chemical Lactic Acid, Racemic, USP is used as a food preservative, curing agent, and flavoring agent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

50-21-5
C3H6O3
UN3265
CC(O)C(O)=O
90.08
USP
0.05%
100%
MFCD00004520
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
2-hydroxypropanoic acid
88 to 92%
4 L
Amber Glass Bottle

Chemical Identifiers

50-21-5
90.08
JVTAAEKCZFNVCJ-UHFFFAOYNA-N
CC(O)C(O)=O
C3H6O3
MFCD00004520
2-hydroxypropanoic acid
Videos
SDS
Documents

Documents

Promotions

Promotions