missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lactose Standard 5%, Apura™ MilliporeSigma™
Supplier: Technidata 1129390010
Specifications
10 g | |
Solid water standard | |
Plastic bottle | |
5% | |
Milk sugar, Lactobiose | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O | |
360.312 | |
360.32 (molar mass) |
Lactose Standard | |
10039-26-6 | |
Solid | |
C12H24O12 | |
WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate | |
104938 |
Chemical Identifiers
10039-26-6 | |
360.312 | |
Milk sugar, Lactobiose | |
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate |
C12H24O12 | |
WSVLPVUVIUVCRA-KPKNDVKVSA-N | |
104938 | |
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O |