missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, Powder, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L124725GM

 View more versions of this product

Catalog No. 18611188


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4618-18-2
Amber Glass Bottle
0.99
1
25 g
Reagent

Chemical Identifiers

4618-18-2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.