National Safety Apparel™ Ladies Coverall 7 oz. Navy Ultrasoft Zipper Closure 32 in. Inseam
For enhanced protection Westex™ UltraSoft is the premium choice for superior comfort, improved safety and above average durability. With a unique blend of 88% cotton and 12% high-tenacity nylon UltraSoft garments provide a smooth feel for increased comfort and the inherently FR material delivers exceptional performance in reliability
$447.35 - $595.42
Specifications
Material Cotton, Nylon
Color Navy
Ankle Style Open
Closure Type Covered Zipper
Seam Type Stitched
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17001493
View Documents
National Safety Apparel™
C88UJWSM32
Small
Each for $447.35
17001494
View Documents
National Safety Apparel™
C88UJWMD32
Medium
Each for $447.35
17001495
View Documents
National Safety Apparel™
C88UJWLG32
Large
Each for $447.35
17001496
View Documents
National Safety Apparel™
C88UJWXL32
X-Large
Each for $447.35
17001497
View Documents
National Safety Apparel™
C88UJW2X32
2X-Large
Each for $492.08
17001498
View Documents
National Safety Apparel™
C88UJW3X32
3X-Large
Each for $492.08
17001499
View Documents
National Safety Apparel™
C88UJW4X32
4X-Large
Each for $541.31
17001500
View Documents
National Safety Apparel™
C88UJW5X32
5X-Large
Each for $595.42
Description
Description
  • Arc Rating: 8 cal/cm²
  • CAT 2
  • Fabric/Material: 7 oz. Westex™ UltraSoft™
  • Comfortable sewn-in elastic waist for better fit
  • Dual certified to protect against flash fire and arc flash hazards
  • 2-way covered zipper front for easy entry
  • Two front chest pockets with flaps and hidden snap closures
  • Large reinforced back pockets
  • Convenient 6 pocket style with side pass through access
  • Leg openings made to fit over work boots
  • All metal covered to ensure no contact with skin
  • Action back allows for full range of motion
  • FR fabric will not melt, drip, or ignite
  • Home laundry
  • Made in the USA
Specifications
Specifications
Cotton, Nylon
Open
Stitched
None
Navy
Covered Zipper
Arc Rating: 8 cal/cm², CAT 2, NFPA 70 E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'