Learn More
Thermo Scientific™ Lambda Exonuclease (10 U/μL)
Lambda Exonuclease is a highly processive exodeoxyribonuclease that selectively digests the 5'-phosphorylated strand of double-stranded DNA in 5'-3' direction.
$141.39 - $567.73
Spécifications
| Concentration | 10 U/μL |
|---|---|
| Enzyme | Exonuclease |
| Product Type | Exonuclease |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
FEREN0561
|
Thermo Scientific™
EN0561 |
1,000 units |
chaque for $141.39
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
FEREN0562
|
Thermo Scientific™
EN0562 |
5,000 units |
chaque for $567.73
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Thermo Scientific Lambda Exonuclease is a highly processive 5'≥3' exodeoxyribonuclease. It selectively digests the 5'-phosphorylated strand of double-stranded DNA. The enzyme exhibits low activity on single-stranded DNA and non-phosphorylated DNA, and has no activity at nicks and limited activity at gaps in DNA.
Highlights
- Active in Thermo Scientific PCR buffers
Spécifications
| 10 U/μL | |
| Exonuclease | |
| Exonuclease |
For Research Use Only. Not for use in diagnostic procedures.