missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lamotrigine, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 460850010

Catalog No. AC460850010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Lamotrigine
0.5% max. (105°C, 3 h)
97.5% min. (HPLC)
C9H7Cl2N5
1g
Solubility in water: insoluble. Other solubilities: soluble in dmf
NC1=NN=C(C(N)=N1)C1=CC=CC(Cl)=C1Cl
256.09
CHEBI:6367
98%
84057-84-1
Authentic
Glass Bottle
MFCD00865333
lamotrigine, 6-2,3-dichlorophenyl-1,2,4-triazine-3,5-diamine, lamictal, lamictal cd, lamotrigina, lamotriginum, lamictal xr, lamotriginum latin, lamotrigina spanish, lamictal odt
PYZRQGJRPPTADH-UHFFFAOYSA-N
6-(2,3-dichlorophenyl)-1,2,4-triazine-3,5-diamine
3878
256.09
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 281-901-8

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only