LaMotte™ Carbon Dioxide Test Kit
Standard alkali is used to titrate samples to the phenolphthalein endpoint.
Supplier:  LaMotte™ 7297DRCN01
Catalog No. 36-103-0805
Explore available promotions
Description
Description
  • Range: 0 - 50ppm
  • Number of tests: 50
Specifications
Specifications
To measure free carbon dioxide in water
50
0 to 50ppm
Carbon Dioxide Test Kit