LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
For water quality testing
Supplier:  LaMotte™ 2991G
 View more versions of this product
Catalog No. 03-920-402
Explore available promotions
Description
Description
  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Specifications
Water quality testing
25 Strips/Vial
Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Copper
0-0.3-0.6-1-3 ppm
Single Factor Test Strip