LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
For water quality testing
Supplier:  LaMotte™ LAM2994
 View more versions of this product
Catalog No. 03-920-403
Explore available promotions
Description
Description
  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Specifications
Water quality testing
25 Tests/Factor/Vial
Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Iron and Copper
0 to 5 ppm (iron), 0 to 3 ppm (copper)
Multi-Factor Test Strip