LaMotte™ Insta-Test™ Single and Multi-Factor Test Strips
For water quality testing
Supplier:  LaMotte™ LAM3002
 View more versions of this product
Catalog No. 22-307-872
Explore available promotions
Description
Description
  • No reagents or field kits required for most strips
  • Chemically treated pads on plastic strips produce color change
  • Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
  • As little as 10-second development time
Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
†Strips have been evaluated for use in these applications. Use in other applications is subject to potential interferences.
Specifications
Specifications
Water quality testing
50 Strips/Vial
Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight
Chlorine dioxide
0 to 500 ppm
Single Factor Test Strip
Product Suggestions
Product Suggestions