LaMotte™ QAC Individual Test Kits
Direct Reading Titrator method allows for precise, convenient QAC testing
Supplier:  LaMotte™ 3043DR
Catalog No. 03-920-386
Explore available promotions
Includes:
  • 15mL QAC Indicator Solution
  • 60 mL QAC Titration Solution
  • One Test Tube, plastic, w/cap
  • One Direct Reading Titrator, 0 to 500 Range
  • One Brush, Test Tube
Description
Description
  • Tests for Quaternary Ammonium Compounds (QACs), such as Alkyl dimethyl benzyl ammonium chloride
  • Range and sensitivity is 0 to 500 ppm in 10 ppm increments
  • Indicator is bromphenol blue
A masked bromphenol blue indicator is added to the sample and turns green. Sodium tetraphenylboron is added to complex the QAC and the color changes to red. This method is best suited to higher QAC concentrations.
For QAC testing in industrial water boiling and cooling, water and wastewater, laundry and sanitation.
Specifications
Specifications
QAC testing in industrial water boiling and cooling, water and wastewater, laundry and sanitation
50
BPB direct reading titrator
15mL QAC indicator solution, 60mL QAC titration solution, 1 test tube, plastic with cap, 1 direct reading titrator, 0 to 500 range, 1 brush, test tube
10 to 500ppm
QAC Test Kit
Videos
SDS
Documents
Documents
Promotions
Promotions
Read the LaMotte Direct Reading Titrator Manual before proceeding. The Titrator is calibrated in parts per million (ppm) QAC; each minor division equals 10 ppm.