missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lanthanum foil, 0.1mm (0.004 in.) thick, 99.9% (REO), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 041558FF

 View more versions of this product

Catalog No. AA41558FF


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gray to Silver
0.1mm (0.004 in.) thick
(REO)
MFCD00011066
25 x 25 mm
FZLIPJUXYLNCLC-UHFFFAOYSA-N
lanthanum
23926
99.9%
≈0.39g/25x25mm
Lanthanum foil
7439-91-0
La
UN3208
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
[La]
138.91
CHEBI:33336
Mylar under Argon
Foil

Chemical Identifiers

7439-91-0
138.91
FZLIPJUXYLNCLC-UHFFFAOYSA-N
23926
lanthanum
La
MFCD00011066
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation
CHEBI:33336
[La]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

EINECSNumber : 231-099-0

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only