Learn More
Lanthanum(III) 2-ethylhexanoate, 10% w/v in hexane, Thermo Scientific Chemicals
$841.96 - $841.96
Chemical Identifiers
CAS | 67816-09-5 |
---|---|
Molecular Formula | C24H45LaO6 |
Molecular Weight (g/mol) | 568.523 |
MDL Number | MFCD00798555 |
InChI Key | PPNFILUQDVDXDA-UHFFFAOYSA-K |
Synonym | lanthanum tris 2-ethylhexanoate, lanthanum 2-ethylhexoate, lanthanum 3+ 2-ethylhexanoate, lanthanum 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lanthanum 3+ salt, hexanoic acid, 2-ethyl-, lanthanum 3+ salt 3:1, 2-ethylhexanoate; lanthanum 3+, tris 2-ethylhexanoic acid lanthanum salt, lanthanum 3+ tris 2-ethylhexanoate |
PubChem CID | 171899 |
IUPAC Name | 2-ethylhexanoate;lanthanum(3+) |
SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[La+3] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA41268AE
|
Thermo Scientific Chemicals
041268AE |
100 mL |
Each for $841.96
|
|
|||||
Description
Lanthanum(III) 2-ethylhexanoate, 10% w/v in hexane is used as drier in ink, coatings and paint industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67816-09-5 | |
568.523 | |
PPNFILUQDVDXDA-UHFFFAOYSA-K | |
171899 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[La+3] |
C24H45LaO6 | |
MFCD00798555 | |
lanthanum tris 2-ethylhexanoate, lanthanum 2-ethylhexoate, lanthanum 3+ 2-ethylhexanoate, lanthanum 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lanthanum 3+ salt, hexanoic acid, 2-ethyl-, lanthanum 3+ salt 3:1, 2-ethylhexanoate; lanthanum 3+, tris 2-ethylhexanoic acid lanthanum salt, lanthanum 3+ tris 2-ethylhexanoate | |
2-ethylhexanoate;lanthanum(3+) |
Specifications
67816-09-5 | |
MFCD00798555 | |
UN1208 | |
Soluble In Organic Solvents. Not miscible in water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[La+3] | |
568.523 | |
568.18 | |
Liquid |
C24H45LaO6 | |
100 mL | |
lanthanum tris 2-ethylhexanoate, lanthanum 2-ethylhexoate, lanthanum 3+ 2-ethylhexanoate, lanthanum 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lanthanum 3+ salt, hexanoic acid, 2-ethyl-, lanthanum 3+ salt 3:1, 2-ethylhexanoate; lanthanum 3+, tris 2-ethylhexanoic acid lanthanum salt, lanthanum 3+ tris 2-ethylhexanoate | |
PPNFILUQDVDXDA-UHFFFAOYSA-K | |
2-ethylhexanoate;lanthanum(3+) | |
171899 | |
10% w/v in hexane | |
Lanthanum(III) 2-ethylhexanoate |
Safety and Handling
GHS H Statement
H225-H304-H361-H373-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
May cause drowsiness or dizziness.
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361f-H373
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXANES
EINECSNumber : 267-212-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only