Learn More
Lanthanum nickel powder, -100 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
$153.49 - $551.01
Chemical Identifiers
CAS | 12196-72-4 |
---|---|
Molecular Formula | LaNi5 |
Molecular Weight (g/mol) | 432.37 |
MDL Number | MFCD00198177 |
InChI Key | BUCVQDCWULHLJZ-UHFFFAOYSA-N |
Synonym | lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy |
PubChem CID | 14367487 |
IUPAC Name | lanthanum;nickel |
SMILES | [Ni].[Ni].[Ni].[Ni].[Ni].[La] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
12196-72-4 | |
432.37 | |
BUCVQDCWULHLJZ-UHFFFAOYSA-N | |
14367487 | |
[Ni].[Ni].[Ni].[Ni].[Ni].[La] |
LaNi5 | |
MFCD00198177 | |
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy | |
lanthanum;nickel |
Specifications
12196-72-4 | |
MFCD00198177 | |
5 g | |
BUCVQDCWULHLJZ-UHFFFAOYSA-N | |
lanthanum;nickel | |
14367487 | |
−100 Mesh | |
Air and moisture sensitive | |
(metals basis) |
LaNi5 | |
UN3178 | |
lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy | |
[Ni].[Ni].[Ni].[Ni].[Ni].[La] | |
432.37 | |
99.5% | |
Odorless | |
Lanthanum nickel powder |
Safety and Handling
GHS H Statement
H228-H260-H350-H317
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P210-P240-P241-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c
H228-H334-H350-H372
DOTInformation : Hazard Class: 4.1; Packaging Group: III
EINECSNumber : 235-372-5
TSCA : Yes
storageNote1 :
- Packaged under Argon
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only