Learn More
Lanthanum rod, 12.7mm (0.5in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
$224.62 - $1106.66
Chemical Identifiers
CAS | 7439-91-0 |
---|---|
Molecular Formula | La |
Molecular Weight (g/mol) | 138.91 |
MDL Number | MFCD00011066 |
InChI Key | FZLIPJUXYLNCLC-UHFFFAOYSA-N |
Synonym | in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation |
PubChem CID | 23926 |
ChEBI | CHEBI:33336 |
IUPAC Name | lanthanum |
SMILES | [La] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA40293HF
|
Thermo Scientific Chemicals
040293HF |
10 mm |
Each for $224.62
|
|
|||||
AA40293DH
|
Thermo Scientific Chemicals
040293DH |
25 mm |
Each for $365.23
|
|
|||||
AA40293DJ
|
Thermo Scientific Chemicals
040293DJ |
50 mm |
Each for $650.12
|
|
|||||
AA40293DM
|
Thermo Scientific Chemicals
040293DM |
100 mm |
Each for $1,106.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Chemical Identifiers
7439-91-0 | |
138.91 | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
23926 | |
lanthanum |
La | |
MFCD00011066 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
CHEBI:33336 | |
[La] |
Specifications
Silver to Gray | |
La | |
UN3208 | |
in chloride, ion 2, lanthanum, ion la2, atom, ingot, pieces, lanthanum, pieces, 2+ ion, foil, 3n, ii cation | |
[La] | |
138.91 | |
CHEBI:33336 | |
Mylar under Argon | |
Cast Rod | |
(metals basis excluding Ta) |
7439-91-0 | |
MFCD00011066 | |
10 mm | |
FZLIPJUXYLNCLC-UHFFFAOYSA-N | |
lanthanum | |
23926 | |
99.9% | |
≈0.80g/mm | |
Lanthanum rod, 12.7mm (0.5 in.) dia. |
Safety and Handling
GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
EINECSNumber : 231-099-0
TSCA : Yes
storageNote1 :
- Packaged under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only