Learn More
Labconco Laptop Arm
Articulating Laptop Arm allows independent placement of a laptop for ease of use while working in a enclosure.
Supplier:  Labconco 3997400
Catalog No. 17-200-205
Only null left
Explore available promotions
Description
Description
  • Articulating Laptop Arm for use with Labconco PC Mount kit and Labconco base stands only
  • Polished Aluminum
  • Allows independent placement of the laptop
  • Capacity 7 to 12 lbs. (3.2 to 5.4 kg)
  • PC Mount kit required and sold separately
  • Installation required
Specifications
Specifications
Enclosures
Laptop Arm
Product Suggestions
Product Suggestions