Learn More
Stoelting™ Large Probe Holder
Stoelting Large Probe Holder supports shafts with diameters from 6.5 mm up to 13 mm (0.5 inch)
Supplier: Stoelting™ 51633
Description
- Ideal for Hamilton™ Micro-syringes or the Stoelting Stereotaxic Flexible Shaft Drill.
- Threaded drive pulls the sliding U holder against the outside of the probe, pulling the probe into a V notch for straight and reproducible positioning.
Specifications
| Neuroscience | |
| Hamilton™ Micro-syringes or Stoelting Stereotaxic Flexible Shaft Drills |