Large Rubber Bulbs
Supplier:  Customer Channels Group (Ccg) 20035EMD
 View more versions of this product
Catalog No. S33300
Explore available promotions
Description
Description
Heavy-walled necks provide secure leak-proof grip
Use these easy to grip rubber bulbs for large pipettes and syringes.
Specifications
Specifications
Not Autoclavable
Rubber
30 mL
2.5 in.
0.25 in.
38 mm
Red
Bulb
Large pipettes and syringes
66 mm
1.5 in.