Justrite™ Large Steel Drum Funnel With Self-Closing Cover
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Large Steel Drum Funnel With Self-Closing Cover helps to prevent spills when transferring flammable solvents, thinners, oils, or paint wastes. This self-closing model meets NFPA1 and the International Fire Code. It shuts and locks automatically when a fusible link melts at 165°F (74°C) under fire conditions.
Supplier:  Justrite™ 08206
 View more versions of this product
Catalog No. 19-822-454
Explore available promotions
Description
Description
  • Justrite’s steel laboratory safety cans have all the benefits of the Type I cans
  • Added features make these the choice of lab professionals for safely dispensing hazardous liquids into test tubes, small flasks and beakers
  • Fill spouts have self-close, pressure-relief caps and stainless steel flame arresters
  • These cans also feature a vacuum-relief valve within the lid for smooth liquid flow, and self-closing faucets for trouble-free dispensing
Specifications
Specifications
Large Steel Drum Funnel
Flammables
32 in. flame arrester with 2 in. drum bung and self-closing cover
Yellow
Steel (Galvanized)