missing translation for 'onlineSavingsMsg'
Learn More

Lauric Anhydride 98.0+%, TCI America™

$44.73 - $607.68

Chemical Identifiers

CAS 645-66-9
Molecular Formula C24H46O3
Molecular Weight (g/mol) 382.629
MDL Number MFCD00008965
InChI Key NWADXBLMWHFGGU-UHFFFAOYSA-N
Synonym lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
PubChem CID 69527
IUPAC Name dodecanoyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L001725G
View Documents
TCI America
L001725G
25 g
Each for $44.73
Only null left
 
L0017100G
View Documents
TCI America
L0017100G
100 g
Each for $123.03
Only null left
 
L0017500G
View Documents
TCI America
L0017500G
500 g
Each for $607.68
Only null left
 
Description

Description

Specifications

Chemical Identifiers

645-66-9
382.629
NWADXBLMWHFGGU-UHFFFAOYSA-N
69527
CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC
C24H46O3
MFCD00008965
lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
dodecanoyl dodecanoate

Specifications

645-66-9
White
MFCD00008965
lauric anhydride, dodecanoic anhydride, lauroyl anhydride, dodecanoic acid, anhydride, n-dodecanoic anhydride, lauricanhydride, dodecanoyl anhydride, lauric acid anhydride, dodecanoic anhydride #, dodecanoicacid,anhydride
CCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCC
382.629
382.63
Crystalline Powder
42°C
C24H46O3
25 g
NWADXBLMWHFGGU-UHFFFAOYSA-N
dodecanoyl dodecanoate
69527
≥98.0% (T)
Lauric Anhydride
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'