missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lauroyl Chloride 98.0+%, TCI America™
$57.27 - $154.05
Chemical Identifiers
CAS | 112-16-3 |
---|---|
Molecular Formula | C12H23ClO |
Molecular Weight (g/mol) | 218.77 |
MDL Number | MFCD00000740 |
InChI Key | NQGIJDNPUZEBRU-UHFFFAOYSA-N |
Synonym | lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 |
PubChem CID | 8166 |
IUPAC Name | dodecanoyl chloride |
SMILES | CCCCCCCCCCCC(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
D097225ML
|
TCI America
D097225ML |
25 mL |
Each for $57.27
|
|
|||||
D0972500ML
|
TCI America
D0972500ML |
500 mL |
Each for $154.05
|
|
|||||
Chemical Identifiers
112-16-3 | |
218.77 | |
NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
8166 | |
CCCCCCCCCCCC(Cl)=O |
C12H23ClO | |
MFCD00000740 | |
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
dodecanoyl chloride |
Specifications
112-16-3 | |
145°C | |
MFCD00000740 | |
3265 | |
NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
dodecanoyl chloride | |
8166 | |
≥98.0% (GC) | |
Lauroyl Chloride |
Yellow | |
C12H23ClO | |
25 mL | |
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
CCCCCCCCCCCC(Cl)=O | |
218.77 | |
218.77 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0632
TSCA : Yes