missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ LB-Agar, Large Capsules
Contents/liter: LB Medium, 15g Agar-B
Supplier: MP Biomedicals™ 113002216
Description
Use 8 large capsules/L.
Specifications
Molecular Biology Certified™ FDA | |
Capsule | |
Agar | |
For growth of bacteria |
15°C to 30°C | |
Molecular Biology | |
Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L | |
227 g |