missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lead Chromate, Spectrum™ Chemical
$2363.29 - $12080.35
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
18611222
|
Spectrum Chemical Mfg Cor
L118325KG |
2.5 kg | Poly Pail |
Each for $2,363.29
|
|
|||||
18611221
|
Spectrum Chemical Mfg Cor
L118312KGBL |
12 kg | Poly Pail |
Each for $8,544.79
|
|
|||||
18611223
|
Spectrum Chemical Mfg Cor
L118350KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | ||||||||||
Chemical Identifiers
7758-97-6 |
Specifications
7758-97-6 | |
2.5 kg | |
Poly Pail |
1 | |
Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.