missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead Dioxide, Powder, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L1063100GM

 View more versions of this product

Catalog No. 18611228


Explore available promotions

Description

Description

Specifications

Specifications

1309-60-0
100 g
Reagent
1
97%
Poly Bottle

Chemical Identifiers

1309-60-0
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.