missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) 2-ethylhexanoate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03671118

 View more versions of this product

Catalog No. AA3671118


Explore available promotions

Description

Description

Films of lead (II) 2-ethylhexanote can be cast on silicon solutions by spin coating. Lead (II) 2-ethylhexanote is a common solvent in metal-organic deposition (MOD) process. Intermetallic PtPb nanoparticles have been synthesized by the chemical reduction of dimethyl(1,5-cyclooctadiene)platinum and lead(II)2-ethylhexanoate by sodium naphthalide in THF or diglyme. The precursors used for the photochemical production of PZT or PLZT films were lead(II) 2-ethylhexanoate, lanthanum(III) 2-ethylhexanoate, zirconyl(IV) 2-ethylhexa- noate, and titanium(IV) isopropoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Lead(II) 2-ethylhexanoate
1.56 g/mL
C16H30O4Pb
50 g
lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+
RUCPTXWJYHGABR-UHFFFAOYSA-L
2-ethylhexanoate;lead(2+)
160451
Viscous Liquid
301-08-6
Odorless
MFCD00014003
UN3082
Insoluble in water.
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2]
493.612
493.61

Chemical Identifiers

301-08-6
493.612
RUCPTXWJYHGABR-UHFFFAOYSA-L
160451
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2]
C16H30O4Pb
MFCD00014003
lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+
2-ethylhexanoate;lead(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H302+H332-H350-H360Df-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

EINECSNumber : 206-107-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only